Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA-PKcs Ab-3, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and gel supershift experiments.

Manufacturer: thermo scientific™ lab vision™  MS371P1ABX

 View more versions of this product

Catalog No. MS371P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen DNA-PKcs Ab-3
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 42-psc; same as 42-26 or 42-27
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen Human DNA-dependent Protein Kinase purified from HeLa cells
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and gel supershift experiments with Thermo Scientific™ DNA-PKcs Ab-3, Mouse Monoclonal Antibody.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 42-psc; same as 42-26 or 42-27

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 2713-3198

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: Testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO