Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ DNA-PKcs Ab-1, Mouse Monoclonal Antibody

Inhibit DNA-dependent phosphorylation and perform other studies with Thermo Scientific™ DNA-PKcs Ab-1, Mouse Monoclonal Antibody.

$271.53 - $1,571.61

Specifications

Antigen DNA-PKcs Ab-1
Clone 18-2
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS369B

 
thermo scientific™ lab vision™
MS369B
Biotinylated Ab with BSA and Azide 1mL Each for $1,571.61

MS369B0

 
thermo scientific™ lab vision™
MS369B0
Biotinylated Ab with BSA and Azide 100μL Each for $360.39

MS369B1

 
thermo scientific™ lab vision™
MS369B1
Biotinylated Ab with BSA and Azide 500μL Each for $991.33

MS369P

 
thermo scientific™ lab vision™
MS369P
Purified Ab with BSA and Azide 1mL Each for $1,257.37

MS369P0

 
thermo scientific™ lab vision™
MS369P0
Purified Ab with BSA and Azide 100μL Each for $271.53

MS369P1

 
thermo scientific™ lab vision™
MS369P1
Purified Ab with BSA and Azide 500μL Each for $753.10

MS369P1ABX

 
thermo scientific™ lab vision™
MS369P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $823.59

MS369PABX

 
thermo scientific™ lab vision™
MS369PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,343.59

MS369R7

 
thermo scientific™ lab vision™
MS369R7
Ready-to-use for IHC (UVLP) 7mL Each for $809.18
Description & Specifications

Specifications

Antigen DNA-PKcs Ab-1
Clone 18-2
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Host Species Murine
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Human DNA-dependent Protein Kinase purified from HeLa cells
Isotype IgG1

Inhibit DNA-dependent phosphorylation and perform other studies with Thermo Scientific™ DNA-PKcs Ab-1, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 18-2

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-2713

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Inhibits (50%) DNA-dependent Phosphorylation

Note:

USA: RUO; Int'l: RUO