Please login to your online account to display your discounted pricing

DMSO, sterile filtered, Tocris

$113.57 - $265.15

Chemical Identifiers

Chemical Name or Material DMSO, sterile filtered
Assay Percent Range >99.9%
Molecular Formula C2H6OS
Formula Weight 78.13
Synonym Dimethyl sulfoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

3176100ML

 
r & d systems
RDS3176100ML
100mL Each for $265.15

317625ML

 
r & d systems
RDS317625ML
25mL Each for $113.57
Description & Specifications

Specifications

Formula Weight 78.13
Quantity 100mL
Chemical Name or Material DMSO, sterile filtered
Assay Percent Range >99.9%
Molecular Formula C2H6OS
Synonym Dimethyl sulfoxide
CAS 67-68-5
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature