Please login to your online account to display your discounted pricing

DMPQ dihydrochloride, Tocris

Potent, selective inhibitor of PDGFRβ

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material DMPQ dihydrochloride
Assay Percent Range >99%
Molecular Formula C16H14N2O2·2HCl
Formula Weight 339.22
Synonym 5, 7-Dimethoxy-3-(4-pyridinyl)quinoline dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

122250

 
r & d systems
122250
50mg Each for $746.48

122210

 
r & d systems
122210
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 339.22
Quantity 50mg
CAS 1123491-15-5
Assay Percent Range >99%
Synonym 5, 7-Dimethoxy-3-(4-pyridinyl)quinoline dihydrochloride
Molecular Formula C16H14N2O2·2HCl
Chemical Name or Material DMPQ dihydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature