Please login to your online account to display your discounted pricing

DMPQ dihydrochloride, Tocris


Potent, selective inhibitor of PDGFRβ

Quantity 10mg

Manufacturer: r & d systems  122210

 View more versions of this product

Catalog No. 122210

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 339.22
Quantity 10mg
CAS 1123491-15-5
Chemical Name or Material DMPQ dihydrochloride
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Molecular Formula C16H14N2O2·2HCl
Synonym 5, 7-Dimethoxy-3-(4-pyridinyl)quinoline dihydrochloride

Specifications

Chemical Name or Material DMPQ dihydrochloride
Assay Percent Range >99%
Molecular Formula C16H14N2O2·2HCl
Formula Weight 339.22
Synonym 5, 7-Dimethoxy-3-(4-pyridinyl)quinoline dihydrochloride
Accessories

Store at Room Temperature