Invitrogen™ DMPK Recombinant Human Protein DFS Item
DMPK is a serine/threonine kinase expressed in various muscle and nerve tissues.
Manufacturer:  Invitrogen™ LSPV3996
 View more versions of this product
Catalog No. PV3996
Description
Description
Mutations in the untranslated region of the DMPK gene are a characteristic of myotonic dystrophy.
Specifications
Specifications
NP_004400
Frozen
96.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
DMPK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
DMPK, DM
Baculovirus
Human
GST Tag
Documents