missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DMNB, Tocris

DNA-PK inhibitor

$75.35 - $257.85

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C9H9NO5
Formula Weight 211.15
Synonym 4, 5-Dimethoxy-2-nitrobenzaldehyde
Chemical Name or Material DMNB
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
208850 R & D Systems
RDS208850
50mg Each for $257.85
208810 R & D Systems
RDS208810
10mg Each for $75.35
Specifications Description & Specifications

Chemical Identifiers

>98%
211.15
DMNB
C9H9NO5
4, 5-Dimethoxy-2-nitrobenzaldehyde

Specifications

50mg
>98%
211.15
DMNB
20357-25-9
C9H9NO5
4, 5-Dimethoxy-2-nitrobenzaldehyde

Safety and Handling

Recommended Storage : Store at -20°C