Please login to your online account to display your discounted pricing

DMH-1, Tocris™

BMP inhibitor; selectively inhibits ALK2

$251.85 - $1,045.45

Chemical Identifiers

Chemical Name or Material DMH-1
Assay Percent Range >98%
Molecular Formula C24H20N4O
Formula Weight 380.44
Synonym 4-[6-[4-(1-Methylethoxy)phenyl]pyrazolo[1, 5-a]pyrimidin-3-yl]-quinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

412610

 
r & d systems
RDS412610
10mg Each for $251.85

412650

 
r & d systems
RDS412650
50mg Each for $1,045.45
Description & Specifications

Specifications

Formula Weight 380.44
Quantity 10mg
Chemical Name or Material DMH-1
Assay Percent Range >98%
Molecular Formula C24H20N4O
CAS 1206711-16-1
Synonym 4-[6-[4-(1-Methylethoxy)phenyl]pyrazolo[1, 5-a]pyrimidin-3-yl]-quinoline
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C