Please login to your online account to display your discounted pricing

DMeOB, Tocris

Negative allosteric modulator at mGlu5

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material DMeOB
Assay Percent Range >98%
Molecular Formula C16H16N2O2
Formula Weight 268.31
Synonym [(3-Methoxyphenyl)methylene]hydrazone-3-methoxybenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

195350

 
r & d systems
RDS195350
50mg Each for $546.19

195310

 
r & d systems
RDS195310
10mg Each for $129.05
Description & Specifications

Specifications

Formula Weight 268.31
Quantity 50mg
Molecular Formula C16H16N2O2
Synonym [(3-Methoxyphenyl)methylene]hydrazone-3-methoxybenzaldehyde
Chemical Name or Material DMeOB
Assay Percent Range >98%
CAS 40252-74-2
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature