missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ DMEM (Dulbecco’s Modified Eagle’s Medium)

Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME

$736.76 - $1002.27

Specifications

Content And Storage 2°C to 8°C
Product Type DMEM Media
Endotoxin Level <0.25 EU/mL
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MT15013LX
View Documents Promotion Details
Corning™
15013LX
10 L
Each for $736.76
 
MT15013LB
View Documents Promotion Details
Corning™
15013LB
20 L
Each for $1,002.27
 
Description

Description

  • Low endotoxin - <0.25 EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
Specifications

Specifications

2°C to 8°C
DMEM Media
<0.25 EU/mL
SDS
missing translation for 'documents'

missing translation for 'documents'