Please login to your online account to display your discounted pricing

DMAB-anabaseine dihydrochloride, Tocris

Partial agonist at α7-containing receptors and antagonist at α4β2 neuronal nicotinic receptors

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material DMAB-anabaseine dihydrochloride
Assay Percent Range >98%
Molecular Formula C19H21N3·2HCl
Formula Weight 364.32
Synonym 4-[(5,6-Dihydro[2,3'-bipyridin]-3(4H)-ylidene)methyl]-N,N-dimethylbenzenamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

224110

 
r & d systems
RDS224110
10mg Each for $225.77

224150

 
r & d systems
RDS224150
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 364.32
Quantity 10mg
Chemical Name or Material DMAB-anabaseine dihydrochloride
Assay Percent Range >98%
Molecular Formula C19H21N3·2HCl
CAS 154149-38-9
Synonym 4-[(5,6-Dihydro[2,3'-bipyridin]-3(4H)-ylidene)methyl]-N,N-dimethylbenzenamine dihydrochloride
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C