Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Tryptophanamide, 98%

Chemical Identifiers

Chemical Name or Material DL-Tryptophanamide
CAS 6720-02-1
Formula Weight 203.24
MDL Number MFCD03826622
Synonym (±)-2-Amino-3-(3-indolyl)propionamide; H-DL-Trp-NH2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6322703

  SDS
alfa aesar™
H6322703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6322706

  SDS
alfa aesar™
H6322706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63227MD

  SDS
alfa aesar™
H63227MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.24
Chemical Name or Material DL-Tryptophanamide
MDL Number MFCD03826622
CAS 6720-02-1
Color Off-white
Quantity 1g
Physical Form Crystalline Powder
Assay 98%
Synonym (±)-2-Amino-3-(3-indolyl)propionamide; H-DL-Trp-NH2