Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Tryptophan methyl ester hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material DL-Tryptophan methyl ester hydrochloride
CAS 5619-09-0
Formula Weight 268.74
MDL Number MFCD00066133
Synonym H-DL-Trp-OMe.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6665906

 
alfa aesar™
H6665906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6665914

 
alfa aesar™
H6665914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 221° to 222°C
Formula Weight 268.74
Chemical Name or Material DL-Tryptophan methyl ester hydrochloride
MDL Number MFCD00066133
Assay 97%
CAS 5619-09-0
Synonym H-DL-Trp-OMe.HCl
Color Off-white to white
Quantity 5g
Physical Form Solid