Learn More
DL-Tropic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2204009
Description
It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.
Solubility
Soluble in water (20 g/L) at 20°C, ethanol, ether, alcohol, and methanol (0.1 g/ml).
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
DL-Tropic acid | |
117°C to 119°C | |
MFCD00004255 | |
2209199 | |
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid | |
JACRWUWPXAESPB-UHFFFAOYNA-N | |
3-hydroxy-2-phenylpropanoic acid | |
10726 | |
166.18 |
552-63-6 | |
C9H10O3 | |
10 g | |
14,9779 | |
Soluble in water (20g/L) at 20°C,ethanol,ether,alcohol,and methanol (0.1g/ml). | |
OCC(C(O)=O)C1=CC=CC=C1 | |
166.18 | |
CHEBI:30765 | |
98% |
Chemical Identifiers
552-63-6 | |
166.18 | |
JACRWUWPXAESPB-UHFFFAOYNA-N | |
10726 | |
3-hydroxy-2-phenylpropanoic acid |
C9H10O3 | |
MFCD00004255 | |
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid | |
CHEBI:30765 | |
OCC(C(O)=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-020-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only