Please login to your online account to display your discounted pricing

DL-TBOA, Tocris

Selective non-transportable inhibitor of EAATs

$264.88 - $1,099.17

Chemical Identifiers

Chemical Name or Material DL-TBOA
Assay Percent Range >98%
Molecular Formula C11H13NO5
Formula Weight 239.23
Synonym DL-threo-β-Benzyloxyaspartic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

122310

 
r & d systems
RDS122310
10mg Each for $264.88

122350

 
r & d systems
RDS122350
50mg Each for $1,099.17
Description & Specifications

Specifications

Formula Weight 239.23
Quantity 10mg
Chemical Name or Material DL-TBOA
Assay Percent Range >98%
Molecular Formula C11H13NO5
CAS 205309-81-5
Synonym DL-threo-β-Benzyloxyaspartic acid
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C