Please login to your online account to display your discounted pricing

DL-TBOA, Tocris


Selective non-transportable inhibitor of EAATs

Quantity 10mg

Manufacturer: r & d systems  RDS122310

 View more versions of this product

Catalog No. 122310

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 239.23
Quantity 10mg
Chemical Name or Material DL-TBOA
Molecular Formula C11H13NO5
Synonym DL-threo-β-Benzyloxyaspartic acid
Assay Percent Range >98%
CAS 205309-81-5
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material DL-TBOA
Assay Percent Range >98%
Molecular Formula C11H13NO5
Formula Weight 239.23
Synonym DL-threo-β-Benzyloxyaspartic acid
Accessories

Desiccate at -20°C