Learn More
DL-sec-Phenethyl alcohol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 351881000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
DL-sec-Phenethyl alcohol | |
98-85-1 | |
Colorless | |
203°C to 205°C | |
Authentic | |
Glass bottle | |
1.5260 to 1.528 | |
MFCD00004508 | |
06, II, 444 | |
methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene | |
WAPNOHKVXSQRPX-UHFFFAOYSA-N | |
1-phenylethanol | |
7409 | |
CHEBI:669 | |
97% |
97% | |
20°C | |
1.0100g/mL | |
86°C | |
96% min. (GC) | |
C8H10O | |
C6H5CH(CH3)OH | |
100 g | |
1.01 | |
Solubility in water: 29g/L (20°C). Other solubilities: soluble in alcohol and glycerol | |
CC(C1=CC=CC=C1)O | |
122.17 | |
11.2 mPa.s (20°C) | |
122.17 | |
Liquid |
Chemical Identifiers
98-85-1 | |
122.17 | |
WAPNOHKVXSQRPX-UHFFFAOYSA-N | |
7409 | |
1-phenylethanol |
C8H10O | |
MFCD00004508 | |
methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene | |
CHEBI:669 | |
CC(C1=CC=CC=C1)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 202-707-1
RTECSNumber : DO9275000
TSCA : TSCA
RUO – Research Use Only