missing translation for 'onlineSavingsMsg'
Learn More

DL-Pyroglutamic Acid 99.0+%, TCI America™

Catalog No. G0061500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0061500G 500 g
G006125G 25 g
G0061100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0061500G Supplier TCI America Supplier No. G0061500G
Only null left

Chemical Identifiers

CAS 149-87-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00064322
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid
PubChem CID 499
ChEBI CHEBI:16010
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1

Specifications

Melting Point 184°C
Color White
Quantity 500 g
Formula Weight 129.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Pyroglutamic Acid
EINECSNumber (5)-0115
TSCA No