missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Pipecolinic Acid 99%, ACROS Organics™

$109.33 - $109.33

Chemical Identifiers

CAS 535-75-1
Assay Percent Range 99%
Molecular Formula C6H11NO2
Formula Weight 129.16
MDL Number MFCD00064347
Synonym DL-2-Piperidinecarboxylic acid
Chemical Name or Material DL-Pipecolinic acid, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC131280250 SDS Thermo Fisher Scientific
131280250
Plastic bottle 25g Each for $109.33
AC131281000 SDS Thermo Fisher Scientific
131281000
Plastic bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

535-75-1
C6H11NO2
MFCD00064347
DL-Pipecolinic acid, 99%
99%
129.16
DL-2-Piperidinecarboxylic acid
≥98.5%

Specifications

Fine Crystalline Powder
268° to 278°C (dec.)
White to light beige
1% max. (K.F.)
Authentic
0.5% max.
535-75-1
MFCD00064347
DL-Pipecolinic acid, 99%
129.16
25g
15, 7570
Plastic bottle
DL-2-Piperidinecarboxylic acid
1% max. (100°C, 4 hrs)
C6H11NO2
99%
≥98.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 223-737-1