Please login to your online account to display your discounted pricing

DL-Pipecolinic Acid 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  131281000

 View more versions of this product

Catalog No. AC131281000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 129.16
Melting Point 268° to 278°C (dec.)
Quantity 100g
Color White to light beige
Merck Index 15, 7570
EINECS Number 223-737-1
Chemical Name or Material DL-Pipecolinic acid
Sulfated Ash 0.5% max.
Water 1% max. (K.F.)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H11NO2
MDL Number MFCD00064347
CAS 535-75-1
Synonym DL-2-Piperidinecarboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Loss on Drying 1% max. (100°C, 4 hrs)
Packaging Plastic bottle

Specifications

Chemical Name or Material DL-Pipecolinic acid
Name Note 99%
CAS 535-75-1
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H11NO2
Formula Weight 129.16
MDL Number MFCD00064347
Synonym DL-2-Piperidinecarboxylic acid
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents