Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Pipecolinic acid, 99%

$113.59 - $304.78

Chemical Identifiers

Chemical Name or Material DL-Pipecolinic acid
CAS 535-75-1
Molecular Formula C6H11NO2
Formula Weight 129.16
MDL Number MFCD00064347
Beilstein 81095
Merck Index 14,7458
Synonym DL-Homoproline; (+/-)-Piperidine-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2354114

 
alfa aesar™
B2354114
25g Each for $113.59

AAB2354122

 
alfa aesar™
B2354122
100g Each for $304.78
Description & Specifications

Specifications

Formula Weight 129.16
Chemical Name or Material DL-Pipecolinic acid
TSCA TSCA
Assay 99%
Molecular Formula C6H11NO2
MDL Number MFCD00064347
CAS 535-75-1
Beilstein 81095
Merck Index 14,7458
Synonym DL-Homoproline; (+/-)-Piperidine-2-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TK6021000
Melting Point ≈280°C (dec.)
Quantity 25g

Applications
DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.

Solubility
Soluble in water (50 mg/ml), and boiling alcohol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.