Learn More
DL-Pipecolinic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2354122
Description
DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.
Solubility
Soluble in water (50 mg/ml), and boiling alcohol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
Specifications
DL-Pipecolinic acid | |
∼280°C (decomposition) | |
MFCD00064347,MFCD00005981 | |
81095 | |
dl-pipecolinic acid, pipecolic acid, pipecolinic acid, 2-piperidinecarboxylic acid, homoproline, pipecolate, dl-pipecolic acid, dihydrobaikiane, piperolinic acid, hexahydropicolinic acid | |
HXEACLLIILLPRG-UHFFFAOYNA-N | |
piperidine-2-carboxylic acid | |
849 | |
129.16 |
535-75-1 | |
C6H11NO2 | |
100 g | |
14,7458 | |
Soluble in water (50mg/ml),and boiling alcohol. | |
OC(=O)C1CCCCN1 | |
129.16 | |
CHEBI:17964 | |
99% |
Chemical Identifiers
535-75-1 | |
129.16 | |
HXEACLLIILLPRG-UHFFFAOYNA-N | |
849 | |
piperidine-2-carboxylic acid |
C6H11NO2 | |
MFCD00064347,MFCD00005981 | |
dl-pipecolinic acid, pipecolic acid, pipecolinic acid, 2-piperidinecarboxylic acid, homoproline, pipecolate, dl-pipecolic acid, dihydrobaikiane, piperolinic acid, hexahydropicolinic acid | |
CHEBI:17964 | |
OC(=O)C1CCCCN1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-616-3
RTECSNumber : TK6021000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only