missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Phenylalanine 98.0+%, TCI America™

Manufacturer:  TCI America P0136250G

 View more versions of this product

Catalog No. P0136250G



Specifications

Specifications

DL-Phenylalanine
250g
C9H11NO2
dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
C1=CC=C(C=C1)CC(C(=O)O)N
165.192
CHEBI:28044
Crystal-Powder at 20°C
White
150-30-1
MFCD00064225
COLNVLDHVKWLRT-UHFFFAOYSA-N
2-amino-3-phenylpropanoic acid
994
165.19
≥98.0% (HPLC,T)

Chemical Identifiers

150-30-1
165.192
COLNVLDHVKWLRT-UHFFFAOYSA-N
994
2-amino-3-phenylpropanoic acid
C9H11NO2
MFCD00064225
dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
CHEBI:28044
C1=CC=C(C=C1)CC(C(=O)O)N
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1283

TSCA : Yes

Documents