missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Phenylalanine, 99%

Manufacturer:  Alfa Aesar™ A1013214

 View more versions of this product

Catalog No. AAA1013214



Description

Description

Applications
DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

Solubility
Soluble in water. Insoluble in ethyl ether, benzene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

DL-Phenylalanine
C9H11NO2
1910407
dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
C1=CC=C(C=C1)CC(C(=O)O)N
165.192
CHEBI:28044
99%
25g
150-30-1
MFCD00064225
14,7271
COLNVLDHVKWLRT-UHFFFAOYSA-N
2-amino-3-phenylpropanoic acid
994
165.19
~266°C (decomposition)

Chemical Identifiers

150-30-1
165.192
COLNVLDHVKWLRT-UHFFFAOYSA-N
994
2-amino-3-phenylpropanoic acid
C9H11NO2
MFCD00064225
dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
CHEBI:28044
C1=CC=C(C=C1)CC(C(=O)O)N
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents