Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Penicillamine, 97+%

$39.70 - $344.80

Chemical Identifiers

Chemical Name or Material DL-Penicillamine
CAS 52-66-4
Molecular Formula C5H11NO2S
Formula Weight 149.21
MDL Number MFCD00004856
Beilstein 2039817
Merck Index 14,7088
Synonym 3,3-Dimethyl-DL-cysteine; 3-Mercapto-DL-valine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2471003

 
alfa aesar™
B2471003
1g Each for $39.70

AAB2471006

 
alfa aesar™
B2471006
5g Each for $82.97

AAB2471014

 
alfa aesar™
B2471014
25g Each for $344.80
Description & Specifications

Specifications

Formula Weight 149.21
MDL Number MFCD00004856
CAS 52-66-4
Recommended Storage 2° to 8°C
Odor Unpleasant
Chemical Name or Material DL-Penicillamine
Assay ≥97%
Molecular Formula C5H11NO2S
Beilstein 2039817
Merck Index 14,7088
Synonym 3,3-Dimethyl-DL-cysteine; 3-Mercapto-DL-valine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number YV9445000
Melting Point ≈212°C (dec.)
Quantity 1g

Applications
Heavy metal chelating agentDL-Penicillamine is used as an antirheumatic. It also serves as a chelating agent for heavy metals. Further, it is used in the treatment of Wilson′s desease, Cystinuria, Scleroderma and arsenic poisoning.

Solubility
Soluble in ethanol, hot water and sodium hydroxide.

Notes
Incompatible with oxidizing agents. Store in a cool place.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C