missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Pantolactone, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 427591000

 View more versions of this product

Catalog No. AC427591000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

DL-Pantolactone
246.0°C
96% min. (GC)
C6H10O3
100g
Solubility in water: soluble.
CC1(COC(=O)C1O)C
130.14
130.14
79-50-5
122°C
Glass bottle
MFCD00064333
pantolactone, dl-pantolactone, 3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, pantoyl lactone, pantoic lactone, dl-pantoyl lactone, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl, dihydro-3-hydroxy-4,4-dimethyl-2 3h-furanone, pantolyl lactone
SERHXTVXHNVDKA-UHFFFAOYSA-N
3-hydroxy-4,4-dimethyloxolan-2-one
989
97%

Chemical Identifiers

79-50-5
130.14
SERHXTVXHNVDKA-UHFFFAOYSA-N
989
CC1(COC(=O)C1O)C
C6H10O3
MFCD00064333
pantolactone, dl-pantolactone, 3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, pantoyl lactone, pantoic lactone, dl-pantoyl lactone, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl, dihydro-3-hydroxy-4,4-dimethyl-2 3h-furanone, pantolyl lactone
3-hydroxy-4,4-dimethyloxolan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 201-210-7

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only