missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-o-Tyrosine, 98%

Manufacturer:  Alfa Aesar™ 4454101

 View more versions of this product

Catalog No. AA4454101



Specifications

Specifications

DL-o-Tyrosine
C9H11NO3
2805789
Soluble in water,and DMSO (Heated).
C1=CC=C(C(=C1)CC(C(=O)O)N)O
181.191
181.19
98%
0.25g
2370-61-8
MFCD00021725
2-amino-3-2-hydroxyphenyl propanoic acid
WRFPVMFCRNYQNR-UHFFFAOYSA-N
2-amino-3-(2-hydroxyphenyl)propanoic acid
91482
Powder
~256°C (decomposition)

Chemical Identifiers

2370-61-8
181.191
WRFPVMFCRNYQNR-UHFFFAOYSA-N
91482
C1=CC=C(C(=C1)CC(C(=O)O)N)O
C9H11NO3
MFCD00021725
2-amino-3-2-hydroxyphenyl propanoic acid
2-amino-3-(2-hydroxyphenyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-134-8

RTECSNumber : YP2285000

TSCA : No

Recommended Storage : Ambient temperatures

Documents