Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-o-Tyrosine, 98%

$98.20 - $310.93

Chemical Identifiers

Chemical Name or Material DL-o-Tyrosine
CAS 2370-61-8
Molecular Formula C9H11NO3
Formula Weight 181.19
MDL Number MFCD00021725
Beilstein 2805789
Synonym 3-(2-Hydroxyphenyl)-DL-alanine; H-DL-Phe(2-OH)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4454101

 
alfa aesar™
4454101
0.25g Each for $98.20

AA4454103

 
alfa aesar™
4454103
1g Each for $310.93
Description & Specifications

Specifications

Formula Weight 181.19
Chemical Name or Material DL-o-Tyrosine
Physical Form Powder
Assay 98%
Molecular Formula C9H11NO3
MDL Number MFCD00021725
CAS 2370-61-8
Beilstein 2805789
Synonym 3-(2-Hydroxyphenyl)-DL-alanine; H-DL-Phe(2-OH)-OH
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈256°C (dec.)
Quantity 0.25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number YP2285000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.