missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-O-Methylserine 98.0+%, TCI America™

Manufacturer:  TCI America M08885G

 View more versions of this product

Catalog No. M08885G



Specifications

Specifications

DL-O-Methylserine
5g
C4H9NO3
KNTFCRCCPLEUQZ-UHFFFAOYSA-N
2-amino-3-methoxypropanoic acid
97963
Crystal-Powder at 20°C
White-Yellow
19794-53-7
MFCD00037773
COCC(C(=O)O)N
119.12
119.12
≥98.0% (T)

Chemical Identifiers

19794-53-7
119.12
KNTFCRCCPLEUQZ-UHFFFAOYSA-N
2-amino-3-methoxypropanoic acid
C4H9NO3
MFCD00037773
97963
COCC(C(=O)O)N
Safety and Handling

Safety and Handling

TSCA : No

Documents