missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-O-Methylserine, TCI America™

$206.25 - $701.27

Chemical Identifiers

CAS 19794-53-7
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00037773
InChI Key KNTFCRCCPLEUQZ-UHFFFAOYSA-N
PubChem CID 97963
IUPAC Name 2-amino-3-methoxypropanoic acid
SMILES COCC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M08885G View Documents TCI America
M08885G
5g Each for $701.27
 
M08881G View Documents TCI America
M08881G
1g Each for $206.25
 
Specifications

Chemical Identifiers

19794-53-7
119.12
KNTFCRCCPLEUQZ-UHFFFAOYSA-N
2-amino-3-methoxypropanoic acid
C4H9NO3
MFCD00037773
97963
COCC(C(=O)O)N

Specifications

White-Yellow
19794-53-7
MFCD00037773
COCC(C(=O)O)N
119.12
119.12
≥98.0% (T)
5g
C4H9NO3
KNTFCRCCPLEUQZ-UHFFFAOYSA-N
2-amino-3-methoxypropanoic acid
97963
Crystal-Powder at 20°C
DL-O-Methylserine
Safety and Handling

Safety and Handling

TSCA : No