Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Nornicotine, 98%

Chemical Identifiers

Chemical Name or Material DL-Nornicotine
CAS 5746-86-1
Molecular Formula C9H12N2
Formula Weight 148.21
MDL Number MFCD00016913
UN Number UN2810
Beilstein 81968
Merck Index 14,6712
Synonym DL-2-(3-Pyridyl)pyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL10809MA

 
alfa aesar™
L10809MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL10809MF

 
alfa aesar™
L10809MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.21
Molecular Formula C9H12N2
Chemical Name or Material DL-Nornicotine
MDL Number MFCD00016913
Assay 98%
CAS 5746-86-1
UN Number UN2810
Beilstein 81968
Merck Index 14,6712
Synonym DL-2-(3-Pyridyl)pyrrolidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number QS6350000
Flash Point 101°C (213°F)
Boiling Point 107° to 108°C (2mmHg)
Quantity 10mg

Applications
DL-Nornicotine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in acetone, chloroform and water (50mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C