missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Nornicotine, 98%

Manufacturer:  Alfa Aesar™ L10809MF

 View more versions of this product

Catalog No. AAL10809MF



Description

Description

Applications
DL-Nornicotine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in acetone, chloroform and water (50mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents.
Specifications

Specifications

DL-Nornicotine
107°C to 108°C (2mmHg)
5746-86-1
MFCD00016913
81968
3-pyrrolidin-2-yl pyridine, nornicotine, 3-2-pyrrolidinyl pyridine, r,s-nornicotine, +--nornicotine, 2-3-pyridyl pyrrolidine, 3-pyrrolidin-2-yl-pyridine, +--1'-demethylnicotine, +/--nornicotine, dl-nornicotine
C1CC(NC1)C2=CN=CC=C2
148.209
148.21
101°C (213°F)
50mg
C9H12N2
UN2810
14,6712
MYKUKUCHPMASKF-UHFFFAOYSA-N
3-pyrrolidin-2-ylpyridine
412
98%

Chemical Identifiers

5746-86-1
148.209
MYKUKUCHPMASKF-UHFFFAOYSA-N
412
C1CC(NC1)C2=CN=CC=C2
C9H12N2
MFCD00016913
3-pyrrolidin-2-yl pyridine, nornicotine, 3-2-pyrrolidinyl pyridine, r,s-nornicotine, +--nornicotine, 2-3-pyridyl pyrrolidine, 3-pyrrolidin-2-yl-pyridine, +--1'-demethylnicotine, +/--nornicotine, dl-nornicotine
3-pyrrolidin-2-ylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : QS6350000

Recommended Storage : 2° to 8°C

Documents