Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Norleucine, 98%

$60.88 - $60.88

Chemical Identifiers

Chemical Name or Material DL-Norleucine
CAS 616-06-8
Molecular Formula C6H13NO2
Formula Weight 131.18
MDL Number MFCD00064422
Beilstein 1721748
Merck Index 14,6706
Synonym (+/-)-2-Aminohexanoic acid; H-DL-Nle-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1079114

  SDS
alfa aesar™
A1079114
25g Each for $60.88

AAA1079122

  SDS
alfa aesar™
A1079122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1079136

  SDS
alfa aesar™
A1079136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material DL-Norleucine
CAS 616-06-8
TSCA TSCA
Assay 98%
Molecular Formula C6H13NO2
Formula Weight 131.18
MDL Number MFCD00064422
Beilstein 1721748
Merck Index 14,6706
Synonym (+/-)-2-Aminohexanoic acid; H-DL-Nle-OH
Melting Point >300°C
Quantity 25g
Odor Odorless