Please login to your online account to display your discounted pricing

DL-N-Benzoyl-2-methylserine, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  339790050

 View more versions of this product

Catalog No. AC339790050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 223.23
Quantity 5g
Color White
Melting Point 153° to 159°C
Infrared Spectrum Authentic
MDL Number MFCD01863306
Name Note 98%
Chemical Name or Material DL-N-Benzoyl-2-methylserine
CAS 7508-82-9
Molecular Formula C11H13NO4
Assay Percent Range 98%
Packaging Glass bottle
Synonym DL-2-Benzoylamino-3-hydroxy-2-methylpropionic acid

Specifications

Chemical Name or Material DL-N-Benzoyl-2-methylserine
Name Note 98%
CAS 7508-82-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H13NO4
Formula Weight 223.23
MDL Number MFCD01863306
Synonym DL-2-Benzoylamino-3-hydroxy-2-methylpropionic acid