missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
DL-N-Benzoyl-2-methylserine, 98%, ACROS Organics™
Chemical Identifiers
CAS |
7508-82-9 |
Molecular Formula |
C11H13NO4 |
Molecular Weight (g/mol) |
223.23 |
MDL Number |
MFCD01863306 |
InChI Key |
FVWQRKJTTRAXJP-NSHDSACASA-N |
Synonym |
2s-2-benzamido-3-hydroxy-2-methylpropanoic acid, 2s-3-hydroxy-2-methyl-2-phenylformamido propanoic acid, n-benzoyl-2-methylserine, s-2-benzamido-3-hydroxy-2-methylpropanoic acid, s-2-methyl-2-benzoylamino-3-hydroxypropanoic acid |
PubChem CID |
7167581 |
IUPAC Name |
(2S)-2-benzamido-3-hydroxy-2-methylpropanoic acid |
SMILES |
CC(CO)(C(=O)O)NC(=O)C1=CC=CC=C1 |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
7508-82-9 |
|
223.23 |
|
FVWQRKJTTRAXJP-NSHDSACASA-N |
|
7167581 |
|
CC(CO)(C(=O)O)NC(=O)C1=CC=CC=C1 |
|
C11H13NO4 |
|
MFCD01863306 |
|
2s-2-benzamido-3-hydroxy-2-methylpropanoic acid, 2s-3-hydroxy-2-methyl-2-phenylformamido propanoic acid, n-benzoyl-2-methylserine, s-2-benzamido-3-hydroxy-2-methylpropanoic acid, s-2-methyl-2-benzoylamino-3-hydroxypropanoic acid |
|
(2S)-2-benzamido-3-hydroxy-2-methylpropanoic acid |
Specifications
|
Authentic |
|
White |
|
1g |
|
98% |
|
MFCD01863306 |
|
Solubility in water: .. Other solubilities: soluble in ethanol,aq. naoh,aq. na2co3 |
|
CC(CO)(C(=O)O)NC(=O)C1=CC=CC=C1 |
|
223.23 |
|
223.23 |
|
98% |
|
Glass bottle |
|
153.0°C to 159.0°C |
|
7508-82-9 |
|
C11H13NO4 |
|
2s-2-benzamido-3-hydroxy-2-methylpropanoic acid, 2s-3-hydroxy-2-methyl-2-phenylformamido propanoic acid, n-benzoyl-2-methylserine, s-2-benzamido-3-hydroxy-2-methylpropanoic acid, s-2-methyl-2-benzoylamino-3-hydroxypropanoic acid |
|
FVWQRKJTTRAXJP-NSHDSACASA-N |
|
(2S)-2-benzamido-3-hydroxy-2-methylpropanoic acid |
|
7167581 |
|
Crystalline Powder |
|
DL-N-Benzoyl-2-methylserine, 98% |
SDS