Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-N-Acetylhomocysteine thiolactone, 99%

$38.18 - $340.18

Chemical Identifiers

Chemical Name or Material DL-N-Acetylhomocysteine thiolactone
CAS 1195-16-0
Molecular Formula C6H9NO2S
Formula Weight 159.21
MDL Number MFCD00005480
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2274106

 
alfa aesar™
B2274106
5g Each for $38.18

AAB2274114

 
alfa aesar™
B2274114
25g Each for $111.28

AAB2274122

 
alfa aesar™
B2274122
100g Each for $340.18
Description & Specifications

Specifications

Formula Weight 159.21
Chemical Name or Material DL-N-Acetylhomocysteine thiolactone
TSCA TSCA
Assay 99%
Molecular Formula C6H9NO2S
MDL Number MFCD00005480
CAS 1195-16-0
RTECS Number AC8680000
Odor Odorless
Melting Point 109° to 111°C
Quantity 5g