Please login to your online account to display your discounted pricing

DL-α-(Methylaminomethyl)benzyl alcohol, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  198230010

 View more versions of this product

Catalog No. AC198230010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Needles
Formula Weight 151.21
Quantity 1g
Color White
Merck Index 10, 4485
RTECS Number DA4976000
Melting Point 74° to 76°C
Chemical Name or Material DL-α-(Methylaminomethyl)benzyl alcohol
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H13NO
Linear Formula C6H5CH(CH2NHCH3)OH
MDL Number MFCD00004506
CAS 6589-55-5
Synonym 2-Methylamino-1-phenylethanol
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material DL-α-(Methylaminomethyl)benzyl alcohol
Name Note 99%
CAS 6589-55-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H13NO
Linear Formula C6H5CH(CH2NHCH3)OH
Formula Weight 151.21
MDL Number MFCD00004506
Synonym 2-Methylamino-1-phenylethanol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents