Please login to your online account to display your discounted pricing

DL-Methyl 2-bromobutyrate, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  217691000

 View more versions of this product

Catalog No. AC217691000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 181.03
Quantity 100g
Color Clear slightly yellow
Density 1.5700g/mL
Beilstein 02, 282
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material DL-Methyl 2-bromobutyrate
Molecular Formula C5H9BrO2
CAS 3196-15-4
MDL Number MFCD00009666
Assay Percent Range 97%
Name Note 97%
Linear Formula C2H5CH(Br)CO2CH3

Specifications

Chemical Name or Material DL-Methyl 2-bromobutyrate
Name Note 97%
CAS 3196-15-4
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H9BrO2
Linear Formula C2H5CH(Br)CO2CH3
Formula Weight 181.03
MDL Number MFCD00009666
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents