missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Methionine Methylsulfonium Chloride, TCI America™

$73.33 - $522.52

Chemical Identifiers

CAS 3493-12-7
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00031700
InChI Key MYGVPKMVGSXPCQ-UHFFFAOYNA-N
Synonym (3-Amino-3-carboxypropyl)dimethylsulfonium Chloride, S-Methylmethioninesulfonium Chloride, Vitamin U Chloride
PubChem CID 14220
IUPAC Name (3-amino-3-carboxypropyl)-dimethylsulfanium;chloride
SMILES C[S+](C)CCC(C(=O)O)N.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M064425G View Documents TCI America
M064425G
25g Each for $73.33
 
M0644500G View Documents TCI America
M0644500G
500g Each for $522.52
 
Specifications

Chemical Identifiers

3493-12-7
199.69
MYGVPKMVGSXPCQ-UHFFFAOYNA-N
14220
C[S+](C)CCC(C(=O)O)N.[Cl-]
C6H14ClNO2S
MFCD00031700
(3-Amino-3-carboxypropyl)dimethylsulfonium Chloride, S-Methylmethioninesulfonium Chloride, Vitamin U Chloride
(3-amino-3-carboxypropyl)-dimethylsulfanium;chloride

Specifications

White
3493-12-7
MFCD00031700
MYGVPKMVGSXPCQ-UHFFFAOYNA-N
(3-amino-3-carboxypropyl)-dimethylsulfanium;chloride
14220
Crystal-Powder at 20°C
DL-Methionine Methylsulfonium Chloride
25g
C6H14ClNO2S
(3-Amino-3-carboxypropyl)dimethylsulfonium Chloride, S-Methylmethioninesulfonium Chloride, Vitamin U Chloride
C[S+](C)CCC(C(=O)O)N.[Cl-]
199.69
199.69
≥99.0% (T)
Safety and Handling

Safety and Handling

TSCA : Yes