Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Methionine ethyl ester hydrochloride, 98+%

$61.03 - $423.41

Chemical Identifiers

Chemical Name or Material DL-Methionine ethyl ester hydrochloride
CAS 6297-53-6
Molecular Formula C7H15NO2S·HCl
Formula Weight 213.73
MDL Number MFCD00237859
Synonym H-DL-Met-OEt·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2578106

  SDS
alfa aesar™
H2578106
5g Each for $61.03

AAH2578114

  SDS
alfa aesar™
H2578114
25g Each for $174.78

AAH2578122

  SDS
alfa aesar™
H2578122
100g Each for $423.41
Description & Specifications

Specifications

Formula Weight 213.73
Quantity 5g
Recommended Storage 2° to 8°C
Melting Point 72° to 75°C
Synonym H-DL-Met-OEt·HCl
CAS 6297-53-6
Molecular Formula C7H15NO2S·HCl
MDL Number MFCD00237859
Assay ≥98%
Chemical Name or Material DL-Methionine ethyl ester hydrochloride
Safety and Handling

2° to 8°C