Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Menthyl borate, 97%

$38.95 - $121.30

Chemical Identifiers

Chemical Name or Material DL-Menthyl borate
CAS 62697-74-9
Molecular Formula C30H57BO3
Formula Weight 476.58
MDL Number MFCD00041735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4370903

 
alfa aesar™
4370903
1g Each for $38.95

AA4370906

 
alfa aesar™
4370906
5g Each for $121.30
Description & Specifications

Specifications

Formula Weight 476.58
Chemical Name or Material DL-Menthyl borate
TSCA TSCA
Physical Form Crystalline
Assay 97%
Molecular Formula C30H57BO3
MDL Number MFCD00041735
CAS 62697-74-9
Solubility Information Soluble in chloroform, ether
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.