missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Malic Acid, TCI America™

Manufacturer:  TCI America M002025G

 View more versions of this product

Catalog No. M002025G



Specifications

Specifications

DL-Malic Acid
C4H6O5
malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
C(C(C(=O)O)O)C(=O)O
134.087
CHEBI:6650
Crystal-Powder at 20°C
130°C
25g
6915-15-7
MFCD00064212
BJEPYKJPYRNKOW-UHFFFAOYSA-N
2-hydroxybutanedioic acid
525
134.09
≥99.0% (T)
White

Chemical Identifiers

6915-15-7
134.087
BJEPYKJPYRNKOW-UHFFFAOYSA-N
525
2-hydroxybutanedioic acid
C4H6O5
MFCD00064212
malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
CHEBI:6650
C(C(C(=O)O)O)C(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1442

RTECSNumber : ON7175000

TSCA : Yes

Documents