missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Malic acid, +99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC125255000

 View more versions of this product

Catalog No. AC125255000



Description

Description

Specifications

Specifications

DL-Malic acid
99+%
HO2CCH2CH(OH)CO2H
2-hydroxysuccinic acid, butanedioic acid, hydroxy, deoxytetraric acid, dl-malic acid, hydroxybutanedioic acid, hydroxysuccinic acid, kyselina jablecna, malate, malic acid, pomalus acid
C(C(C(=O)O)O)C(=O)O
134.087
CHEBI:6650
99+%
Solubility in water: 558g/L (20°C). Other solubilities: 82.70 g/100 g methanol-17.75 g/100 g acetone (20°C, 45.53 g/100 g ethanol-20.70 g/ 100 g dioxane (20°C, 0.84 g/100 g diethyl ether (20°C), practically insoluble in benzene
500g
6915-15-7
C4H6O5
MFCD00064212
BJEPYKJPYRNKOW-UHFFFAOYSA-N
2-hydroxybutanedioic acid
525
134.09
Plastic bottle
128.0°C to 132.0°C
203°C

Chemical Identifiers

6915-15-7
134.087
BJEPYKJPYRNKOW-UHFFFAOYSA-N
525
2-hydroxybutanedioic acid
C4H6O5
MFCD00064212
2-hydroxysuccinic acid, butanedioic acid, hydroxy, deoxytetraric acid, dl-malic acid, hydroxybutanedioic acid, hydroxysuccinic acid, kyselina jablecna, malate, malic acid, pomalus acid
CHEBI:6650
C(C(C(=O)O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS P Statement Causes serious eye irritation.

GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

SDS Documents