missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Malic acid, +99%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 125250010
Specifications
DL-Malic acid | |
99+% | |
HO2CCH2CH(OH)CO2H | |
2-hydroxysuccinic acid, butanedioic acid, hydroxy, deoxytetraric acid, dl-malic acid, hydroxybutanedioic acid, hydroxysuccinic acid, kyselina jablecna, malate, malic acid, pomalus acid | |
C(C(C(=O)O)O)C(=O)O | |
134.087 | |
CHEBI:6650 | |
99+% | |
Solubility in water: 558g/L (20°C). Other solubilities: 82.70 g/100 g methanol-17.75 g/100 g acetone (20°C, 45.53 g/100 g ethanol-20.70 g/ 100 g dioxane (20°C, 0.84 g/100 g diethyl ether (20°C), practically insoluble in benzene | |
1kg |
6915-15-7 | |
C4H6O5 | |
MFCD00064212 | |
BJEPYKJPYRNKOW-UHFFFAOYSA-N | |
2-hydroxybutanedioic acid | |
525 | |
134.09 | |
Plastic bottle | |
128.0°C to 132.0°C | |
203°C |
Chemical Identifiers
6915-15-7 | |
134.087 | |
BJEPYKJPYRNKOW-UHFFFAOYSA-N | |
525 | |
2-hydroxybutanedioic acid |
C4H6O5 | |
MFCD00064212 | |
2-hydroxysuccinic acid, butanedioic acid, hydroxy, deoxytetraric acid, dl-malic acid, hydroxybutanedioic acid, hydroxysuccinic acid, kyselina jablecna, malate, malic acid, pomalus acid | |
CHEBI:6650 | |
C(C(C(=O)O)O)C(=O)O |
Safety and Handling
GHS P Statement Causes serious eye irritation.
GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger