missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Leucylglycylglycine, TCI America™

Manufacturer:  TCI America L0033100MG

 View more versions of this product

Catalog No. L0033100MG



Specifications

Specifications

DL-Leucylglycylglycine
100mg
C10H19N3O4
H-DL-Leu-Gly-Gly-OH
CC(C)CC(C(=O)NCC(=O)NCC(=O)O)N
245.279
245.28
≥99.0% (T)
White-Yellow
4337-37-5
MFCD00064990
VWHGTYCRDRBSFI-UHFFFAOYSA-N
2-[[2-[(2-amino-4-methylpentanoyl)amino]acetyl]amino]acetic acid
96806
Crystal-Powder at 20°C

Chemical Identifiers

4337-37-5
245.279
VWHGTYCRDRBSFI-UHFFFAOYSA-N
96806
CC(C)CC(C(=O)NCC(=O)NCC(=O)O)N
C10H19N3O4
MFCD00064990
H-DL-Leu-Gly-Gly-OH
2-[[2-[(2-amino-4-methylpentanoyl)amino]acetyl]amino]acetic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents