missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Leucylglycyl-DL-phenylalanine 95.0+%, TCI America™

Manufacturer:  TCI America L0035100MG

Catalog No. L0035100MG



Specifications

Specifications

DL-Leucylglycyl-DL-phenylalanine
100mg
C17H25N3O4
H-DL-Leu-Gly-DL-Phe-OH
CC(C)CC(C(=O)NCC(=O)NC(CC1=CC=CC=C1)C(=O)O)N
335.404
335.40
≥95.0% (HPLC,N)
White
4294-25-1
MFCD00020428
KEVYYIMVELOXCT-UHFFFAOYSA-N
2-[[2-[(2-amino-4-methylpentanoyl)amino]acetyl]amino]-3-phenylpropanoic acid
274161
Crystal-Powder at 20°C

Chemical Identifiers

4294-25-1
335.404
KEVYYIMVELOXCT-UHFFFAOYSA-N
274161
CC(C)CC(C(=O)NCC(=O)NC(CC1=CC=CC=C1)C(=O)O)N
C17H25N3O4
MFCD00020428
H-DL-Leu-Gly-DL-Phe-OH
2-[[2-[(2-amino-4-methylpentanoyl)amino]acetyl]amino]-3-phenylpropanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents