missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Leucylglycine, TCI America™

$35.14 - $149.72

Chemical Identifiers

CAS 615-82-7
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.227
MDL Number MFCD00021729
InChI Key LESXFEZIFXFIQR-UHFFFAOYSA-N
Synonym H-DL-Leu-Gly-OH
PubChem CID 79070
IUPAC Name 2-[(2-amino-4-methylpentanoyl)amino]acetic acid
SMILES CC(C)CC(C(=O)NCC(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L0030100MG View Documents TCI America
L0030100MG
100mg Each for $35.14
 
L00301G View Documents TCI America
L00301G
1g Each for $149.72
 
Specifications

Chemical Identifiers

615-82-7
188.227
LESXFEZIFXFIQR-UHFFFAOYSA-N
79070
CC(C)CC(C(=O)NCC(=O)O)N
C8H16N2O3
MFCD00021729
H-DL-Leu-Gly-OH
2-[(2-amino-4-methylpentanoyl)amino]acetic acid

Specifications

White
615-82-7
MFCD00021729
LESXFEZIFXFIQR-UHFFFAOYSA-N
2-[(2-amino-4-methylpentanoyl)amino]acetic acid
79070
Crystal-Powder at 20°C
DL-Leucylglycine
100mg
C8H16N2O3
H-DL-Leu-Gly-OH
CC(C)CC(C(=O)NCC(=O)O)N
188.227
188.23
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No