missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 56217-82-4 |
---|---|
Molecular Formula | C15H22N2O3 |
Molecular Weight (g/mol) | 278.352 |
MDL Number | MFCD00037258 |
InChI Key | KFKWRHQBZQICHA-UHFFFAOYSA-N |
Synonym | H-DL-Leu-DL-Phe-OH |
PubChem CID | 259325 |
IUPAC Name | 2-[(2-amino-4-methylpentanoyl)amino]-3-phenylpropanoic acid |
SMILES | CC(C)CC(C(=O)NC(CC1=CC=CC=C1)C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
L0038100MG |
TCI America
L0038100MG |
100mg |
Each for $47.36
|
|
L00381G |
TCI America
L00381G |
1g |
Each for $171.11
|
|
Chemical Identifiers
56217-82-4 | |
278.352 | |
KFKWRHQBZQICHA-UHFFFAOYSA-N | |
259325 | |
CC(C)CC(C(=O)NC(CC1=CC=CC=C1)C(=O)O)N |
C15H22N2O3 | |
MFCD00037258 | |
H-DL-Leu-DL-Phe-OH | |
2-[(2-amino-4-methylpentanoyl)amino]-3-phenylpropanoic acid |
Specifications
White | |
56217-82-4 | |
MFCD00037258 | |
KFKWRHQBZQICHA-UHFFFAOYSA-N | |
2-[(2-amino-4-methylpentanoyl)amino]-3-phenylpropanoic acid | |
259325 | |
Crystal-Powder at 20°C | |
DL-Leucyl-DL-phenylalanine |
100mg | |
C15H22N2O3 | |
H-DL-Leu-DL-Phe-OH | |
CC(C)CC(C(=O)NC(CC1=CC=CC=C1)C(=O)O)N | |
278.352 | |
278.35 | |
≥98.0% (T) |
Safety and Handling
TSCA : No