missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Lactide, TCI America™

Manufacturer:  TCI America L009125G

 View more versions of this product

Catalog No. L009125G



Specifications

Specifications

DL-Lactide
White
25g
C6H8O4
dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
CC1C(=O)OC(C(=O)O1)C
144.126
144.13
≥98.0% (T)
142°C
124°C
95-96-5
MFCD00011685
JJTUDXZGHPGLLC-UHFFFAOYSA-N
3,6-dimethyl-1,4-dioxane-2,5-dione
7272
Crystal-Powder at 20°C

Chemical Identifiers

95-96-5
144.126
JJTUDXZGHPGLLC-UHFFFAOYSA-N
7272
CC1C(=O)OC(C(=O)O1)C
C6H8O4
MFCD00011685
dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
3,6-dimethyl-1,4-dioxane-2,5-dione
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5295

RTECSNumber : JH1100000

TSCA : Yes

Documents