Please login to your online account to display your discounted pricing

DL-Lactic acid, 85+%, ACS reagent, ACROS Organics™


Quantity 2.5kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC412960025

 View more versions of this product

Catalog No. AC412960025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Slightly Viscous Liquid
Formula Weight 90.08
Quantity 2.5kg
Color Clear colorless
Merck Index 15, 5384
Assay 85.0 to 90.0%
TSCA TSCA
RTECS Number OD2800000
EINECS Number 200-018-0
Density 1.2000g/mL
Specific Rotation Condition −0.05 (25.00°C c= neat)
Residue after Ignition 0.02% max.
Iron (Fe) 5ppm max.
Heavy Metals (as Pb) 5ppm max.
CAS 7732-18-5
MDL Number MFCD00004520
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Substances Darkened by H2SO4 Passes Test
Chlorine (Cl) 10ppm max.
Synonym DL-2-Hydroxypropanoic acid
Sulfate 20ppm max.
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye damage.
Chemical Name or Material DL-Lactic acid
CAS 50-21-5
Molecular Formula C3H6O3
Linear Formula CH3CH(OH)CO2H
Assay Percent Range 85+%

Specifications

Chemical Name or Material DL-Lactic acid
CAS 50-21-5
CAS 7732-18-5
Purity Grade ACS Reagent Grade
Assay 85.0 to 90.0%
Assay Percent Range 85+%
Molecular Formula C3H6O3
Linear Formula CH3CH(OH)CO2H
Formula Weight 90.08
MDL Number MFCD00004520
Synonym DL-2-Hydroxypropanoic acid
Accessories

Danger

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye damage.

SDS & Safety Documents